Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KEKEMDPFH YDYQTLRIGG LVFAVVLFSV GILLILSRRC KCSFNQKPRA PGDEEAQVEN LITANATEPQ KAEN (23-95 aa encoded by BC018652)
This gene encodes a member of the FXYD family of transmembrane proteins. This particular protein encodes phosphohippolin, which likely affects the activity of Na,K-ATPase. Multiple alternatively spliced transcript variants encoding the same protein have been described. Related pseudogenes have been identified on chromosomes 10 and X. Read-through transcripts have been observed between this locus and the downstream sodium/potassium-transporting ATPase subunit gamma (FXYD2, GeneID 486) locus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FXYD domain-containing ion transport regulator 6; Phosphohippolin; PLM-like protein; Vascular endothelial cell-specific protein 6; VESP6
Gene Aliases: 0610030I18Rik; FXYD6; Php; Plp; UNQ521/PRO1056
UniProt ID: (Human) Q9H0Q3, (Mouse) Q9D164, (Rat) Q91XV6
Entrez Gene ID: (Human) 53826, (Mouse) 59095, (Rat) 63847
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support