Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MGCVQCKDKE ATKLTEERDG SLNQSSGYRY GTDPTPQHYP SFGVTSIPNY NNFHAAGGQG LTVFGGVNSS SHTGTLRTRG GTGVTLFVAL YDYEARTEDD LSFHKGEKFQ ILNSSEGDWW EARSLTTGET GYIPSNYVAP VDSIQAEEWY FGKLGRKDAE RQLLSFGNPR GTFLIRESET TKGAYSLSIR DWDDMKGDHV KHYKIRKLDN GGYYITTRAQ FETLQQLVQH YSGTWNGNTK VAIKTLKPGT MSPESFLEEA QIMKKLKHDK LVQLYAVVSE EPIYIVTEYM NKGSLLDFLK DGEGRALKLP NLVDMAAQVA AGMAYIERMN YIHRDLRSAN ILVGNGLICK IADFGLARLI EDNEYTARQG AKFPIKWTAP EAALYGRFTI KSDVWSFGIL LTELVTKGRV PYPGMNNREV LEQVERGYRM PCPQDCPISL HELMIHCWKK DPEERPTFEY LQSFLEDYFT ATEPQYQPGE NL
FYN is a SRC-related kinase expressed in brain, skeletal muscle and adipose tissues. Fyn functions in a large number of pathways and has been shown to play a role in memory. FYN is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C syn protooncogene; c-syn protooncogene; EC 2.7.10.2; FYN oncogene related to SRC, FGR, YES; kinase Fyn; MGC45350; OKT3-induced calcium influx regulator; OTTHUMP00000017916; OTTHUMP00000017917; P59 p59-Fyn; p59-Fyn; Proto-oncogene c-Fyn; Proto-oncogene Syn; Protooncogene Syn; RP1-66H14.1; SLK; Src like kinase; Src-like kinase; src/yes-related novel; tyrosine kinase p59fyn(T); Tyrosine-protein kinase Fyn
Gene Aliases: FYN; p59-FYN; SLK; SYN
UniProt ID: (Human) P06241
Entrez Gene ID: (Human) 2534
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support