Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: GAGEICVGQN TSDGSGGPGG GPTAYPTAPY LPDLPFTALP PGASDGRGRP AFPFSCPRQL KVPPYLGYRF LGERDCGAPC EPGRANGLMY FKEEERRFA
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD7 protein contains an N-terminal signal sequence, 10 cysteine residues typical of the cysteine-rich extracellular domain of Fz family members, 7 putative transmembrane domains, and an intracellular C-terminal tail with a PDZ domain-binding motif. FZD7 gene expression may downregulate APC function and enhance beta-catenin-mediated signals in poorly differentiated human esophageal carcinomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: frizzled 7, seven transmembrane spanning receptor; frizzled family receptor 7; frizzled homolog 7; Frizzled, drosophila, homolog of, 7; Frizzled-7; Fz-7; FzE3; hFz7; rizzled 7
Gene Aliases: FZD7; FzE3
UniProt ID: (Human) O75084
Entrez Gene ID: (Human) 8324
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support