Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Hemostasis following tissue injury involves the deployment of essential plasma procoagulants which are involved in a blood coagulation cascade leading to the formation of insoluble fibrin clots and the promotion of platelet aggregation. Factor XII, (FXII) a blood coagulation factor, is a serum glycoprotein that participates in fibrinolysis, as well as the generation of bradykinin and angiotensin. An enzyme of the serine protease (or serine endopeptidase) class, it activates both Factor XI and prekallikrein in the coagulation cascade. Factor XII deficiency, a rare hereditary disorder slightly more prevalent among Asians, does not cause excessive hemorrhaging since other coagulation factors compensate for it. Researchers have still reported Factor XII deficiency to be a risk factor for the development of arterial and venous thromboembolism. The gene for human Factor XII maps to the very end of the long arm of the fifth chromosome (5q33-qter).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-factor XIIa part 1; beta-factor XIIa part 2; Coagulation factor XII; coagulation factor XII (Hageman factor); coagulation factor XIIa heavy chain; coagulation factor XIIa light chain; Factor 12; factor XII; Factor12; HAF; Hageman factor
Gene Aliases: F12; FXII; HAE3; HAEX; HAF
UniProt ID: (Human) P00748, (Rat) D3ZTE0, (Mouse) Q80YC5
Entrez Gene ID: (Human) 2161, (Rat) 306761, (Mouse) 58992
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support