Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Members of the F-box protein family, such as FBXL12, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F-box and leucine-rich repeat protein 12; F-box protein containing leucine repeats 12; F-box protein FBL12; F-box/LRR-repeat protein 12; FLJ20188
Gene Aliases: 3110048D16Rik; FBL12; FBXL12
UniProt ID: (Human) Q9NXK8, (Mouse) Q9EPX5
Entrez Gene ID: (Human) 54850, (Rat) 313782, (Mouse) 30843
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support