Limited time savings – buy 3, only pay for 2

Shop now

Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins and Peptides
  • ELISA Kits
  • SNP Genotyping
  • 所有文檔和支持
  • 證書
  • SDS(安全資料表
  • 產品常見問題
  • Documents & Support
  • 手冊和試驗方法
    Search button Close
            Target of Interest
            Application
            Target Species
            • 訂單查詢
            • 快速訂購
            • 登入
              登入
              還沒有帳戶嗎 ? 立即註冊
              • 帳戶
              • 訂單
              • Connect: Lab, Data, Apps
              • 客製化產品與項目
              • 儀器管理

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Ferritin Light Chain Antibodies

            Invitrogen

            Ferritin Light Chain Polyclonal Antibody

            1 Published Figure
            1 Reference
            View all (50) Ferritin Light Chain antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # PA5-29599
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (TWD)
            18,800.00
            線上優惠
            Ends: 30-Jun-2025
            24,000.00
            Save 5,200.00 (22%)
            100 µL
             
            Check your price & availability
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite Ferritin Light Chain Polyclonal Antibody

            • Antibody Testing Data (4)
            • Published Figures (1)
            Application
            Ferritin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 5

            Ferritin Light Chain Antibody (PA5-29599) in IHC (P)

            Immunohistochemical analysis of paraffin-embedded Ca922 xenograft, using Ferritin Light Chain (Product # PA5-29599) antibody at 1:100 dilution. Antigen Retrieval: EDTA based buffer, pH 8.0, 15 min.
            View Product

            Ferritin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ferritin Light Chain Antibody in Western Blot (WB)
            Ferritin Light Chain Antibody in Western Blot (WB)
            Ferritin Light Chain Antibody in Western Blot (WB)
            Ferritin Light Chain Antibody in Immunocytochemistry (ICC/IF)

            Product Details

            PA5-29599

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:3,000
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:100-1:1,000
            -

            Immunocytochemistry (ICC/IF)

            -
            View 1 publication 1 publication

            ELISA (ELISA)

            Assay-dependent
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Recombinant protein encompassing a sequence within the center region of human Ferritin Light Chain. The exact sequence is proprietary.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            0.1M tris glycine, pH 7, with 10% glycerol

            Contains

            0.01% thimerosal

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2547075

            Product Specific Information

            Recommended positive controls: HepG2, mouse liver.

            Predicted reactivity: Dog (87%), Cat (86%), Pig (88%), Rabbit (86%), Rhesus Monkey (97%), Bovine (84%), Guinea pig (84%).

            Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

            Target Information

            This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.

            ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: Ferritin L subunit; Ferritin L subunit 1; ferritin L-chain; Ferritin light chain; Ferritin light chain 1; ferritin light polypeptide-like 3; ferritin, light polypeptide; fril

            View more View less

            Gene Aliases: FTL; Ftl-1; Ftl1; L-ferritin; LFTD; NBIA3

            View more View less

            UniProt ID: (Human) P02792, (Rat) P02793

            View more View less

            Entrez Gene ID: (Human) 2512, (Mouse) 14325, (Rat) 29292

            View more View less

            Function(s)

            iron ion binding protein binding ferric iron binding identical protein binding molecular_function storage protein

            Process(es)

            iron ion transport cellular iron ion homeostasis iron ion homeostasis response to lead ion
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            產品訂購 Plus Icon Minus Icon
            • 訂單狀態
            • 訂購協助
            • 快速訂購
            • 現貨供應中心
            • 電子採購
            支持 Plus Icon Minus Icon
            • 協助與支持
            • 聯絡我們
            • 技術支持中心
            • 搜尋文件和證書
            • 回報網站問題
            資源 Plus Icon Minus Icon
            • 學習中心
            • 促銷活動
            • 活動和網路研討會
            • 社群媒體
            關於 Thermo Fisher Plus Icon Minus Icon
            • 關於我們 關於我們
            • 人才招募 人才招募
            • 投資者資訊 投資者資訊
            • 新聞 新聞
            • 企業社會責任 企業社會責任
            • 商標
            品牌 Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie偏好

              在此網站使用 Cookie 的選擇

              我們以及我們的附屬公司和供應商使用 cookies 和類似技術來經營我們的網站、識別我們網站的訪客、提供安全登入、收集統計資料以優化網站功能,並提供根據您的興趣量身定制的內容。點擊「全部接受」 以接受所有 cookies,並直接進入網站,或點擊「管理設定」 以查看 cookies 類型的詳細說明,並選擇是否在網站接受某些 cookies。
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Taiwan flag icon
            Taiwan

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0