Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
SLC40A1 is an iron-regulated transporter assumed to play roles in intestinal iron absorption and cellular iron release. It mediates iron efflux in the presence of ferroxidase. Defects in SLC40A1 cause hemochromatosis type 4 (HFE4), which is an autosomal dominant iron-loading disorder.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAR1; Cell adhesion regulator; duodenal-specific; Ferroportin; ferroportin 1; Ferroportin-1; iron regulated gene 1; Iron-regulated transporter 1; Metal transporter protein 1; metal transporting protein 1; MTP1; polycythaemia; SLC11A3 iron transporter; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3; solute carrier family 39 (iron-regulated transporter), member 1; solute carrier family 40 (iron-regulated transporter), member 1; Solute carrier family 40 member 1
Gene Aliases: Dusg; FPN; FPN1; Fpt1; HFE4; IREG1; MST079; MSTP079; MTP; MTP1; Ol5; Pcm; SLC11A3; Slc39a1; SLC40A1
UniProt ID: (Human) Q9NP59, (Mouse) Q9JHI9, (Rat) Q923U9
Entrez Gene ID: (Human) 30061, (Mouse) 53945, (Rat) 170840
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support