Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: ADSGEGDFL AEGGGVRGPR VVERHQSACK DSDWPFCSGE DWNYKCPSGC RMKGLIDEVN QDFTNRINKL KNSLFEYQKN NKDSHSLTTN IMEILRGDFS SANNRDNTYN RVSEDLRSRI EVLKRKVIEK VTANNLLVAR VTTEETPHLK ARAIKWQMRP EVKPIMKEHI APREAMLNLA LSEVSTLLLW GSLPCPPRLS (20-218 aa encoded by BC020764)
Fibrinogen is a soluble protein found in blood plasma and is produced by the liver cells. The protein is a hexamer consisting of two sets of disulfide linked chains. Fibrinogen is essential for blood coagulation as the precursor of fibrin. It is converted to fibrin by the action of thrombin in the presence of calcium ions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fibrinogen a chain; Fibrinogen alpha; Fibrinogen alpha chain; Fibrinogen α chain; fibrinogen, A alpha polypeptide; Fibrinogenα chain
Gene Aliases: ENSMUSG00000059807; FGA; Fib; Fib2
UniProt ID: (Human) P02671, (Mouse) E9PV24
Entrez Gene ID: (Human) 2243, (Mouse) 14161
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support