Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: IQLTYNPDE SSKPNMIDAA TLKSRKMLEE IMKYEASILT HDSSIRYLQE IYNSNNQKIV NLKEKVAQLE AQCQEPCKDT VQIHDITGKD CQDIANKGAK QSGLYFIKPL KANQQFLVYC EIDGSGNGWT VFQKRLDGSV DFKKNWIQYK EGFGHLSPTG TTEFWLGNEK IHLISTQSAI PYALRVELED WNGRTSTADY AMFKVGPEAD KYRLTYAYFA GGDAGDAFDG FDFGDDPSDK FFTSHNGMQF STWDNDNDKF EGNCAEQDGS GWWMNKCHAG HLNGVYYQGG TYSKASTPNG YDNGIIWATW KTRWYSMKKT TMKIIPFNRL TIGEGQQHHL GGAKQAGDV (90-437 aa encoded by BC007044)
The protein encoded by this gene is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibg; Fibrinogen gamma chain; fibrinogen, gamma polypeptide; testicular tissue protein Li 70
Gene Aliases: FGG; PRO2061
UniProt ID: (Human) P02679
Entrez Gene ID: (Human) 2266
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support