Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, HeLa, HepG2, Mouse liver, Rat liver.
Predicted reactivity: Mouse (97%), Rat (95%), Pig (98%), Bovine (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the fibulin family of extracellular matrix glycoproteins. Like all members of this family, the encoded protein contains tandemly repeated epidermal growth factor-like repeats followed by a C-terminus fibulin-type domain. This gene is upregulated in malignant gliomas and may play a role in the aggressive nature of these tumors. Mutations in this gene are associated with Doyne honeycomb retinal dystrophy. Alternatively spliced transcript variants that encode the same protein have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EGF containing fibulin-like extracellular matrix protein 1; EGF-containing fibulin-like extracellular matrix protein 1; Extracellular protein S1-5; FIBL-3; Fibrillin-like protein; Fibulin-3; FLJ35535; MGC111353
Gene Aliases: DHRD; DRAD; EFEMP1; FBLN3; FBNL; FIBL-3; MLVT; MTLV; S1-5
UniProt ID: (Human) Q541U7, (Mouse) Q8BPB5
Entrez Gene ID: (Human) 2202, (Mouse) 216616
Molecular Function:
extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support