Product References
Utilization of ex vivo tissue model to study skin regeneration following microneedle stimuli.
Scientific reports
Liu X,Barresi R,Kaminer M,Qian K,Thillou F,Bataillon M,Liao IC,Zheng Q,Bouez C
Published figure using Filaggrin monoclonal antibody (Product # MA5-13440) in Immunohistochemistry
Thu Oct 27 00:00:00 UTC 2022
FGF21 promotes migration and differentiation of epidermal cells during wound healing via SIRT1-dependent autophagy.
British journal of pharmacology
Chen X,Tong G,Fan J,Shen Y,Wang N,Gong W,Hu Z,Zhu K,Li X,Jin L,Cong W,Xiao J,Zhu Z
MA5-13440 was used in Flow Cytometry, Immunohistochemistry (PFA fixed) to show that FGF21 is a key regulator of keratinocyte migration and differentiation during wound healing. FGF21 may be a novel therapeutic target to accelerate would healing.
Tue Mar 01 00:00:00 UTC 2022
FGF21 promotes migration and differentiation of epidermal cells during wound healing via SIRT1-dependent autophagy.
British journal of pharmacology
Chen X,Tong G,Fan J,Shen Y,Wang N,Gong W,Hu Z,Zhu K,Li X,Jin L,Cong W,Xiao J,Zhu Z
MA5-13440 was used in Flow Cytometry, Immunohistochemistry (PFA fixed) to show that FGF21 is a key regulator of keratinocyte migration and differentiation during wound healing. FGF21 may be a novel therapeutic target to accelerate would healing.
Tue Mar 01 00:00:00 UTC 2022
Human Mesenchymal Stromal Cell-Derived Exosomes Promote In Vitro Wound Healing by Modulating the Biological Properties of Skin Keratinocytes and Fibroblasts and Stimulating Angiogenesis.
International journal of molecular sciences
Tutuianu R,Rosca AM,Iacomi DM,Simionescu M,Titorencu I
MA5-13440 was used in Immunocytochemistry to test if MSC-derived exosomes could accelerate wound healing by enhancing the biological properties of the main cell types involved in the key phases of this process.
Wed Jun 09 00:00:00 UTC 2021
Effect of Fibrin Concentration on the In Vitro Production of Dermo-Epidermal Equivalents.
International journal of molecular sciences
Montero A,Quílez C,Valencia L,Girón P,Jorcano JL,Velasco D
MA5-13440 was used in Immunohistochemistry-immunofluorescence to determine the optimal final fibrin concentration for designing a functional skin dermo-epidermal equivalent for in vitro applications.
Wed Jun 23 00:00:00 UTC 2021
Fully Three-Dimensional Bioprinted Skin Equivalent Constructs with Validated Morphology and Barrier Function.
Tissue engineering. Part C, Methods
Derr K,Zou J,Luo K,Song MJ,Sittampalam GS,Zhou C,Michael S,Ferrer M,Derr P
MA5-13440 was used in Immunohistochemistry to develop a high throughput, reproducible, three-dimensional bioprinted skin equivalents that are morphologically and functionally comparable to native skin tissue is advancing research in skin diseases, and providing a physiologically relevant platform for the development of therapeutics, transplants for regenerative medicine, and testing of skin products like cosmetics.
Sat Jun 01 00:00:00 UTC 2019
Fully Three-Dimensional Bioprinted Skin Equivalent Constructs with Validated Morphology and Barrier Function.
Tissue engineering. Part C, Methods
Derr K,Zou J,Luo K,Song MJ,Sittampalam GS,Zhou C,Michael S,Ferrer M,Derr P
MA5-13440 was used in Immunohistochemistry to develop a high throughput, reproducible, three-dimensional bioprinted skin equivalents that are morphologically and functionally comparable to native skin tissue is advancing research in skin diseases, and providing a physiologically relevant platform for the development of therapeutics, transplants for regenerative medicine, and testing of skin products like cosmetics.
Sat Jun 01 00:00:00 UTC 2019
Saurian papulosis: a new clinicopathological entity.
Journal of the American Academy of Dermatology
Molina-Ruiz AM,del Carmen Fariña M,Carrasco L,Santonja C,Rodríguez-Peralto JL,Torrelo A,Kutzner H,Requena L
MA5-13440 was used in immunohistochemistry to characterize two clinical cases of saurian papulosis
Fri Feb 01 00:00:00 UTC 2013
The regulation of epithelial cell proliferation and growth by IL-1 receptor antagonist.
Biomaterials
Kondo M,Yamato M,Takagi R,Namiki H,Okano T
MA5-13440 was used in immunohistochemistry to study the ability of IL-1 receptor antagonist to regulate the growth and proliferation of epithelial cells and the significance for tissue engineering of epithelial sheets
Tue Jan 01 00:00:00 UTC 2013
Mutations in SERPINB7, encoding a member of the serine protease inhibitor superfamily, cause Nagashima-type palmoplantar keratosis.
American journal of human genetics
Kubo A,Shiohama A,Sasaki T,Nakabayashi K,Kawasaki H,Atsugi T,Sato S,Shimizu A,Mikami S,Tanizaki H,Uchiyama M,Maeda T,Ito T,Sakabe J,Heike T,Okuyama T,Kosaki R,Kosaki K,Kudoh J,Hata K,Umezawa A,Tokura Y,Ishiko A,Niizeki H,Kabashima K,Mitsuhashi Y,Amagai M
MA5-13440 was used in immunohistochemistry to study Nagashima-type palmoplantar keratosis caused by putative loss-of-function mutations in SERPINB7
Thu Nov 07 00:00:00 UTC 2013
Pituitary tumor transforming gene 1 induces tumor necrosis factor-α production from keratinocytes: implication for involvement in the pathophysiology of psoriasis.
The Journal of investigative dermatology
Ishitsuka Y,Kawachi Y,Maruyama H,Taguchi S,Fujisawa Y,Furuta J,Nakamura Y,Ishii Y,Otsuka F
MA5-13440 was used in immunocytochemistry and immunohistochemistry to study the induction of keratinocyte TNF-alpha production by the PTTG1 transcription factor and the significance for psoriasis
Fri Nov 01 00:00:00 UTC 2013
Papillary fibroblasts differentiate into reticular fibroblasts after prolonged in vitro culture.
Experimental dermatology
Janson D,Saintigny G,Mahé C,El Ghalbzouri A
MA5-13440 was used in immunohistochemistry to study the propensity of papillary fibroblasts to develop a reticular fibroblast phenotype following prolonged passage in vitro
Tue Jan 01 00:00:00 UTC 2013
Regulation of epithelial differentiation and proliferation by the stroma: a role for the retinoblastoma protein.
The Journal of investigative dermatology
Pickard A,Cichon AC,Menges C,Patel D,McCance DJ
MA5-13440 was used in immunocytochemistry to study the function of retinoblastoma protein in the stromal compartment
Sat Dec 01 00:00:00 UTC 2012
A decisive function of transforming growth factor-β/Smad signaling in tissue morphogenesis and differentiation of human HaCaT keratinocytes.
Molecular biology of the cell
Buschke S,Stark HJ,Cerezo A,Prätzel-Wunder S,Boehnke K,Kollar J,Langbein L,Heldin CH,Boukamp P
MA5-13440 was used in immunocytochemistry to study the role of TGF-beta/Smad signaling in tissue morphogenesis and differentiation of human HaCaT keratinocytes
Tue Mar 15 00:00:00 UTC 2011
Human embryonic stem cell-derived keratinocytes exhibit an epidermal transcription program and undergo epithelial morphogenesis in engineered tissue constructs.
Tissue engineering. Part A
Metallo CM,Azarin SM,Moses LE,Ji L,de Pablo JJ,Palecek SP
MA5-13440 was used in Immunohistochemistry to study the functional similarity of human embryonic stem cell derivatives and associated primary cell types to validate their utility.
Fri Jan 01 00:00:00 UTC 2010
Establishment and characterization of immortalized human gingival keratinocyte cell lines.
Journal of periodontal research
Gröger S,Michel J,Meyle J
MA5-13440 was used in flow cytometry to establish and characterize the immortalized human gingival keratinocyte cell lines
Mon Dec 01 00:00:00 UTC 2008
Characterization of multipotent cells from human adult hair follicles.
Toxicology in vitro : an international journal published in association with BIBRA
Raposio E,Guida C,Baldelli I,Curto M,Fiocca R,Kunkl A,Robello G,Santi PL
MA5-13440 was used in immunohistochemistry to characterize the multipotent endothelial cells from stripped human hair follicle
Thu Mar 01 00:00:00 UTC 2007
A characteristic subset of psoriasis-associated genes is induced by oncostatin-M in reconstituted epidermis.
The Journal of investigative dermatology
Gazel A,Rosdy M,Bertino B,Tornier C,Sahuc F,Blumenberg M
MA5-13440 was used in immunohistochemistry to study the role of oncostatin-M in inducing a characteristic subset of psoriasis-associated genes in reconstituted epidermis
Fri Dec 01 00:00:00 UTC 2006