Search Thermo Fisher Scientific
- Contact Us
- Quick Order
Invitrogen
FIGURE: 1 / 1
Endotoxin level < 0.01EU/µg by LAL method
Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Fremanezumab is a biosimilar that targets CALCA .