Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Furin Antibodies

            Invitrogen

            Furin Polyclonal Antibody

            Advanced Verification
            31 Published Figures
            54 References
            View all (16) Furin antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # PA1-062
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            612.00
            100 µL
             
            In stock
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite Furin Polyclonal Antibody

            • Antibody Testing Data (6)
            • Published Figures (31)
            • Advanced Verification (4)
            Application
            Furin Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 41

            Furin Antibody (PA1-062) in ICC/IF

            Immunofluorescence analysis of Furin was performed using 70% confluent log phase BeWo cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Furin Polyclonal Antibody (Product # PA1-062) at 2 µg/mL dilution in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor Plus 488 (Product # A327... View More
            View Product

            Furin Antibody in Immunocytochemistry (ICC/IF)
            Furin Antibody in Immunocytochemistry (ICC/IF)
            Furin Antibody in Immunocytochemistry (ICC/IF)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Immunocytochemistry (ICC/IF)
            Furin Antibody in Immunocytochemistry (ICC/IF)
            Furin Antibody in Immunocytochemistry (ICC/IF)
            Furin Antibody in Immunocytochemistry (ICC/IF)
            Furin Antibody in Western Blot, Immunocytochemistry, Immunoprecipitation (WB, ICC/IF, IP)
            Furin Antibody in Immunocytochemistry (ICC/IF)
            Furin Antibody in Immunocytochemistry (ICC/IF)
            Furin Antibody in Immunocytochemistry (ICC/IF)
            Furin Antibody in Immunohistochemistry (IHC)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Western Blot, Immunoprecipitation (WB, IP)
            Furin Antibody in Western Blot, Immunoprecipitation (WB, IP)
            Furin Antibody in Western Blot (WB)
            Furin Antibody in Immunoprecipitation (IP)
            Furin Antibody in Immunoprecipitation (IP)
            Furin Antibody
            Furin Antibody
            Furin Antibody
            Furin Antibody

            Product Details

            PA1-062

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:2,000
            View 22 publications 22 publications

            Immunohistochemistry (IHC)

            -
            View 3 publications 3 publications

            Immunocytochemistry (ICC/IF)

            2 µg/mL
            View 23 publications 23 publications

            Immunoprecipitation (IP)

            1:100
            View 3 publications 3 publications

            Neutralization (Neu)

            -
            View 3 publications 3 publications
            Product Specifications

            Species Reactivity

            Dog, Human, Mouse, Rat

            Published species

            Dog, Hamster, Human, Mouse, Non-human primate, Pig, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Synthetic peptide corresponding to residues R(780) G E R T A F I K D Q S A L(793) of human Furin.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            2 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS with 1mg/mL BSA

            Contains

            0.05% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2105077

            Product Specific Information

            PA1-062 detects furin convertase from canine and mouse cells as well as transfected human furin. This antibody does not detect endogenous furin from BSC-40, HeLa, J774A.1 BPAEC, or CHO cells, or from rat skeletal muscle, spleen, kidney, ovary, testes, heart, or brain tissue.

            PA1-062 has been successfully used in Western blot, immunocytochemistry, immunoprecipitation, and immunofluorescence procedures. By Western blot, this antibody detects an ~100 kDa protein representing furin in BSC-40 cells transfected with the human furin gene. In addition, PA1-062 detects human furin transfected into 3T3 cells. Immunofluorescence staining of endogenous furin convertase in 3T3 cells with PA1-062 yields a pattern consistent with paranuclear staining.

            The PA1-062 immunogen is a synthetic peptide corresponding to residues R(780) G E R T A F I K D Q S A L(793) of human Furin. The PA1-062 immunizing peptide (Cat. # PEP-010) is available for use in neutralization and control experiments.

            Target Information

            Furin is a membrane-associated, calcium-dependent, serine protease that belongs to the subtilisin-like prohormone convertase (PC) family. Members of this family of cellular enzymes cleave most prohormones and neuropeptide precursors. Numerous other cellular proteins, some viral proteins, and bacterial toxins that are transported by the constitutive secretory pathway are also targeted for maturation by PCs. Furin and other PC family members share structural similarities which include a heterogeneous ~10 kDa amino-terminal proregion, a highly conserved ~55 kDa subtilisin-like catalytic domain, and carboxyl-terminal domain that is heterogeneous in length and sequence. These enzymes become catalytically active following proregion cleavage within the appropriate cellular compartment. Furin is the only known PC to possess a transmembrane domain. Cleavage of target proteins occurs at the carboxyl-terminus of the furin consensus sequence, RX(K/R)R. It has been shown that the acidic peptide sequence, C771PSDSEEDEG780, localizes furin to the trans-Golgi-network. Phosphorylation of serine residues within this region modulates intracellular routing of furin protein. An additional signaling domain includes the tetrapeptide sequence, Y759KGL762, which directs internalization from the cell surface.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Bioinformatics

            Protein Aliases: dibasic processing enzyme; Dibasic-processing enzyme; FES upstream region; Furin; furin (paired basic amino acid cleaving enzyme); furin, membrane associated receptor protein; PACE; Paired basic amino acid cleaving enzyme (furin); paired basic amino acid residue cleaving enzyme; Paired basic amino acid residue-cleaving enzyme; Prohormone convertase 3; proprotein convertase subtilisin/kexin type 3; Proprotein convertase subtilisin/kexin type 3 (paired basic amino acid cleaving enzyme, furin, membrane associated receptor protein); proprotein convertase subtilisin/kexin type3; subtilisin pro-protein processing enzyme

            View more View less

            Gene Aliases: 9130404I01Rik; FUR; FURIN; PACE; PCSK3; SPC1

            View more View less

            UniProt ID: (Human) P09958, (Rat) P23377, (Mouse) P23188

            View more View less

            Entrez Gene ID: (Dog) 488746, (Human) 5045, (Rat) 54281, (Mouse) 18550

            View more View less

            Function(s)

            protease binding endopeptidase activity serine-type endopeptidase activity serine-type endopeptidase inhibitor activity protein binding peptidase activity peptide binding metal ion binding nerve growth factor binding serine-type peptidase activity hydrolase activity serine protease

            Process(es)

            signal peptide processing transforming growth factor beta receptor signaling pathway aging cell proliferation negative regulation of endopeptidase activity protein processing peptide hormone processing viral life cycle viral protein processing extracellular matrix disassembly extracellular matrix organization positive regulation of cell migration positive regulation of transforming growth factor beta receptor signaling pathway collagen catabolic process nerve growth factor processing negative regulation of low-density lipoprotein particle receptor catabolic process nerve growth factor production negative regulation of nerve growth factor production negative regulation of transforming growth factor beta1 production secretion by cell regulation of protein catabolic process peptide biosynthetic process cellular protein metabolic process regulation of low-density lipoprotein particle receptor biosynthetic process positive regulation of membrane protein ectodomain proteolysis regulation of endopeptidase activity proteolysis regulation of signal transduction
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-ps2qb:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0