Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epididymis tissue protein Li 17E; G alpha 15; G alpha 16; G alpha-15; G alpha-16; G-protein subunit alpha-15; G-protein subunit alpha-16; GNA 15; GNA 16; Gq class; guanine nucleotide binding protein (G protein), alpha 15 (Gq class); Guanine nucleotide-binding protein subunit alpha-15; Guanine nucleotide-binding protein subunit alpha-16
Gene Aliases: G[a]15; Galpha15; Gna-15; GNA15; GNA16
UniProt ID: (Human) P30679, (Mouse) P30678, (Rat) O88302
Entrez Gene ID: (Human) 2769, (Mouse) 14676, (Rat) 89788
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support