Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
FIGURE: 1 / 2
Reconstitution: 1 X 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. The subunit encoded by this gene is expressed in several non-neuronal tissues including the uterus and ovaries. This subunit can assemble with known GABA A receptor subunits, and the presence of this subunit alters the sensitivity of recombinant receptors to modulatory agents such as pregnanolone. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor subunit pi; GABA-A receptor pi subunit; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, pi; gamma-aminobutyric acid (GABA-A) receptor, pi; gamma-aminobutyric acid A receptor, pi; Gamma-aminobutyric acid receptor subunit pi; MGC126386; MGC126387
Gene Aliases: Gabrp
UniProt ID: (Mouse) Q8QZW7, (Rat) O09028
Entrez Gene ID: (Mouse) 216643, (Rat) 81658
Molecular Function: ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support