Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MTKREAEELI EIEIDGTEKA ECTEESIVEQ TYAPAECVSQ AIDINEPIGN LKKLLEPRLQ CSLDAHEICL QDIQLDPERS LFDQGVKTDG TVQLSVQVIS YQGIEPKLNI LEIVKPADTV EVVIDPDAHH AESEAHLVEE AQVITLDGTK HITTISDETS EQVTRWAAAL EGYRKEQERL GIPYDPIQWS TDQVLHWVVW VMKEFSMTDI DLTTLNISGR ELCSLNQEDF FQRVPRGEIL WSHLELLRKY VLASQEQQMN EIVTIDQPVQ IIPASVQSAT PTTIKVINSS AKAAKVQRAP RISGEDRSSP GNRTGNNGQI QLWQFLLELL TDKDARDCIS WVGDEGEFKL NQPELVAQKW GQRKNKPTMN YEKLSRALRY YYDGDMICKV QGKRFVYKFV CDLKTLIGYS AAELNRLVTE CEQKKLAKMQ LHGIAQPVTA VALSTASLQT EKDN
This gene encodes one of three GA-binding protein transcription factor subunits which functions as a DNA-binding subunit. Since this subunit shares identity with a subunit encoding the nuclear respiratory factor 2 gene, it is likely involved in activation of cytochrome oxidase expression and nuclear control of mitochondrial function. This subunit also shares identity with a subunit constituting the transcription factor E4TF1, responsible for expression of the adenovirus E4 gene. Because of its chromosomal localization and ability to form heterodimers with other polypeptides, this gene may play a role in the Down Syndrome phenotype. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GA binding protein transcription factor alpha subunit 60kDa; GA binding protein transcription factor, alpha subunit 60kDa; GA-binding protein alpha chain; GABP subunit alpha; human nuclear respiratory factor-2 subunit alpha; nuclear respiratory factor 2 alpha subunit; Nuclear respiratory factor 2 subunit alpha; Transcription factor E4TF1-60
Gene Aliases: E4TF1-60; E4TF1A; GABPA; NFT2; NRF2; NRF2A; RCH04A07
UniProt ID: (Human) Q06546
Entrez Gene ID: (Human) 2551
Molecular Function: winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support