Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 11
Positive controls: Rat brain tissue lysates, 293T, LOVO, rat kidney tissue, human skin tissue, human breast tissue, human breast carcinoma tissue, human kidney tissue, mouse kidney tissue, mouse heart tissue, SH-SY5Y.
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor; GABA(A) receptor subunit beta-1; GABA(A) receptor, beta 1; GABA(A)R beta-1; GABA-RB; GABAAR subunit beta-1; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 1; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 1; gamma-aminobutyric acid A receptor beta 1; Gamma-aminobutyric acid receptor beta 1; Gamma-aminobutyric acid receptor subunit beta-1; gamma-aminobutyric acid receptor, subunit beta 1; GBRB1
Gene Aliases: AW061132; B230208N19Rik; Gabrb-1; GABRB1; GARB1
UniProt ID: (Human) P18505, (Mouse) P50571, (Rat) P15431
Entrez Gene ID: (Human) 2560, (Mouse) 14400, (Rat) 25450
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support