Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • GABRB3 Antibodies

            Invitrogen

            GABRB3 Polyclonal Antibody

            View all (24) GABRB3 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # PA5-41056
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (EUR)
            514,00
            100 µL
             
            Check your price
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite GABRB3 Polyclonal Antibody

            • Antibody Testing Data (1)
            Application
            GABRB3 Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            GABRB3 Antibody (PA5-41056) in WB

            Western blot analysis of human liver cells using an anti-GABRB3 polyclonal antibody (Product # PA5-41056).
            View Product

            GABRB3 Antibody in Western Blot (WB)

            Product Details

            PA5-41056

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            0.2-1 µg/mL
            -
            Product Specifications

            Species Reactivity

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Synthetic peptide directed towards the middle region of human GABRB3

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.5 mg/mL

            Purification

            Affinity Chromatography

            Storage buffer

            PBS with 2% sucrose

            Contains

            0.09% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2609058

            Product Specific Information

            Peptide sequence: ESYGYTTDDI EFYWRGGDKA VTGVERIELP QFSIVEHRLV SRNVVFATGA

            Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%

            Target Information

            Gamma-amino butyric acid (GAMA) is a strong inhibitor of neuron transmission and is found throughout the central nervous system. GABA is formed by the decarboxylation of glutamate, resulting in the formation of one of two distinct types; GABA-A or GABA-B.The binding of GABA to GABA-A receptors results in the increase transportation of chlorine ions into the neuron. This influx results in signal transmission repression. Studies have shown that if cell to cell signaling goes unchecked due to a lack of GABA, a system overload can resulting in blackouts, rapid body contractions, seizures and epilepsy.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: cleft palate 1; GABA(A) receptor subunit beta-3; GABA-alpha receptor beta-2 subunit; GABAA receptor beta-3 subunit; GABAAR subunit beta-3; GABR B3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 3; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 3; gamma-aminobutyric acid A receptor beta 3; Gamma-aminobutyric acid receptor subunit beta-3; MGC9051

            View more View less

            Gene Aliases: A230092K12Rik; AW049585; beta3; Cp1; ECA5; Gabrb-3; GABRB3

            View more View less

            UniProt ID: (Human) P28472, (Mouse) P63080

            View more View less

            Entrez Gene ID: (Human) 2562, (Mouse) 14402

            View more View less

            Function(s)

            GABA-A receptor activity GABA-gated chloride ion channel activity ion channel activity extracellular ligand-gated ion channel activity chloride channel activity AP-2 adaptor complex binding ligand-gated ion channel

            Process(es)

            transport signal transduction sensory perception of sound ion transmembrane transport negative regulation of neuron apoptotic process palate development inhibitory postsynaptic potential inner ear receptor cell development innervation cellular response to histamine cochlea development chloride transmembrane transport ion transport chloride transport gamma-aminobutyric acid signaling pathway regulation of neuron apoptotic process neuron development synaptic transmission, GABAergic negative regulation of neuron death
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            France flag icon
            France

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-v98pj:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0