Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Control: HT-29 cell lysates, MCF-7.
Subcellular Location: Postsynaptic cell membrane, cell membrane.
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the gaba/benzodiazepine receptor and opening an integral chloride channel.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor subunit delta; GABA(A) receptor, delta; GABA-A receptor, delta polypeptide; GABAARdelta; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, delta; Gamma-aminobutyric acid receptor subunit delta; MGC105467; MGC45284
Gene Aliases: EIG10; EJM7; GABRD; GEFSP5
UniProt ID: (Human) O14764
Entrez Gene ID: (Human) 2563
Molecular Function: ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support