Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
2 µg/mL of MA5-45484 was sufficient for detection of Delta1 GABA-A receptor in 10 µg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 55kDa.
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the gaba/benzodiazepine receptor and opening an integral chloride channel.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor subunit delta; GABA(A) receptor, delta; GABA-A receptor delta-subunit; GABA-A receptor, delta polypeptide; GABAA receptor delta subunit; GABAARdelta; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, delta; gamma-aminobutyric acid (GABA-A) receptor, subunit delta; gamma-aminobutyric acid A receptor, delta; Gamma-aminobutyric acid receptor subunit delta; MGC105467; MGC45284
Gene Aliases: AI853201; EIG10; EJM7; GABAA-RD; GABRD; GEFSP5
UniProt ID: (Human) O14764, (Mouse) P22933, (Rat) P18506
Entrez Gene ID: (Human) 2563, (Mouse) 14403, (Rat) 29689
Molecular Function: ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support