Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: GABRR2.
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA receptors, which are ligand-gated chloride channels. The protein encoded by this gene is a member of the rho subunit family and is a component of the GABA receptor complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor subunit rho-2; GABA(C) receptor; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) receptor, rho 2; gamma-aminobutyric acid (GABA-A) receptor subunit rho 2; gamma-aminobutyric acid (GABA-A) receptor, subunit rho 2; gamma-aminobutyric acid (GABA-C) receptor, subunit rho 2; gamma-aminobutyric acid A receptor, rho 2; gamma-aminobutyric acid receptor rho2 subunit; Gamma-aminobutyric acid receptor subunit rho-2
Gene Aliases: Gabrr2
UniProt ID: (Rat) P47742, (Mouse) P56476
Entrez Gene ID: (Rat) 29695, (Mouse) 14409
Molecular Function: ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support