Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide sequence: FCCDNDINIV RVSGMQRLAQ LLGEPAETQG TTEARDLHCL LVTNPHTDAW
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%
Cellcycle progression issubject to arrest at G1 and G2 checkpointsin response to DNA damage, presumablyto allow time for DNA repair prior to entryinto S andMphase, respectively. The p53 tumorsuppressor isrequired for one such G1 checkpoint and functionsto upregulate expression of GADD 45 and p21. GADD 45 binds both Cdks and PCNA, a protein involved in DNA replication and repair. GADD 45 stimulates DNA excision repair in vitro and inhibits entry ofcellsinto S phase. Thus, it has been suggested that GADD 45 mayserve as a link between the p53-dependentcellcycle checkpoint and DNA repair. GADD 45-like proteins, GADD 45 beta and GADD 45 gamma, have been shown to be induced by environmentalstresses. GADD 45 beta and GADD 45 gamma are thought to induce p38/JNK activation via MEKK4 activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP566B133; Growth arrest and DNA damage-inducible protein GADD45 beta; growth arrest and DNA-damage-inducible protein GADD45 beta; growth arrest and DNA-damage-inducible, beta; myeloid differentiation primary response gene 118; Myeloid differentiation primary response protein MyD118; Negative growth regulatory protein MyD118
Gene Aliases: AI323528; GADD45B; GADD45BETA; MYD118
UniProt ID: (Human) O75293, (Mouse) P22339
Entrez Gene ID: (Human) 4616, (Mouse) 17873
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support