Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGE5 is a gene that encodes a protein belonging to the GAGE family. The GAGE (G antigen) genes are a group of highly homologous genes that are normally expressed in the testis but are also expressed in various types of cancer. The specific function of GAGE5 is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease. However, studies have suggested that GAGE family proteins may play a role in promoting tumor growth and metastasis, as well as in modulating the immune response to cancer cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 4.4; Cancer/testis antigen 4.5; cancer/testis antigen family 4, member 4; cancer/testis antigen family 4, member 5; CT4.4; CT4.5; G antigen 4; G antigen 5; GAGE-4; GAGE-5; GAGE5; GAGE5_HUMAN
Gene Aliases: CT4.4; CT4.5; GAGE4; GAGE5
UniProt ID: (Human) P0DSO3, (Human) Q13069
Entrez Gene ID: (Human) 2576, (Human) 2577
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support