Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji.
Predicted reactivity: Mouse (84%), Rat (84%), Dog (89%), Pig (89%), Rhesus Monkey (96%), Chimpanzee (99%), Bovine (88%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Catalyzes the sulfation of membrane glycolipids. Seems to prefer beta-glycosides at the non-reducing termini of sugar chains attached to a lipid moiety. Catalyzes the synthesis of galactosylceramide sulfate (sulfatide), a major lipid component of the myelin sheath and of monogalactosylalkylacylglycerol sulfate (seminolipid), present in spermatocytes. Also acts on lactosylceramide, galactosyl 1-alkyl-2-sn-glycerol and galactosyl diacylglycerol (in vitro).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-phosphoadenosine-5'-phosphosulfate:GalCer sulfotransferase; 3'-phosphoadenylylsulfate:galactosylceramide 3'-sulfotransferase; cerebroside (3'-phosphoadenylylsulfate:galactosylceramide 3') sulfotransferase; Cerebroside sulfotransferase; Galactosylceramide sulfotransferase; GalCer sulfotransferase
Gene Aliases: CST; GAL3ST1
UniProt ID: (Human) Q99999
Entrez Gene ID: (Human) 9514
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support