Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: LDAGLYDHF NATFWRHVAR AGRACVEREA RELREARQRL LRRCFGDEPL LRPAAQIRTK QLQPWQPSRK VDIMGYDLPG GGAGPATEAC LKLAMPEVQY SNYLLRKQKR RGGARARPEP VLDNPPPRPI RVLPRGPQGP (293-0 aa encoded by BC113552)
GAL3ST3 is a member of the galactose-3-O-sulfotransferase protein family. It catalyzes sulfonation by transferring a sulfate group to the 3' position of galactose in N-acetyllactosamine in both type 2 (Gal-beta-1-4GlcNAc-R) oligosaccharides and core-2-branched O-glycans, but not on type 1 or core-1-branched structures. This gene, which has also been referred to as GAL3ST2, is different from the GAL3ST2 gene located on chromosome 2 that encodes a related enzyme with distinct tissue distribution and substrate specificities, compared to galactose-3-O-sulfotransferase 3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-galactose-3-O-sulfotransferase 3; Gal-beta-1, 3-GalNAc 3'-sulfotransferase 3; Gal3ST-3; Gal3ST3; galactose 3'-sulfotransferase; Galactose-3-O-sulfotransferase 3; galbeta1-3GalNAc 3'-sulfotransferase 3; MGC142112; MGC142114
Gene Aliases: GAL3ST-3; GAL3ST2; GAL3ST3
UniProt ID: (Human) Q96A11, (Mouse) P61315
Entrez Gene ID: (Human) 89792, (Mouse) 545276, (Rat) 499302
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support