Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Sequence of this protein is as follows: YVLDDSDGLG REFDGIGAVS GGGATSRLLV NYPEPYRSQI LDYLFKPNFG ASLHILKVEI GGDGQTTDGT EPSHMHYALD GNYFRGYEWW LMKEAKKRNP NITLIGLPWS FPGWLGKGFD WPYVNLQLTA YYVVTWIVGA KRYHDLDIDY IGIWNERSYN ANYIKILRKM LNYQGLQRVK IIASDNLWES ISASMLLDAE LFKVVDVIGA HYPGTHSAKD AKLTGKKLWS SEDFSTLNSD MGAGCWGRIL NQNYINGYMT STIAWNLVAS YYEQLPYGRC GLMTAQEPWS GHYVVESPVW VSAHTTQFTQ PGWYYLKTVG HLEKGGSYVA LTDGLGNLTI IIETMSHKHS KCIRPFLPYF NVSQQFATFV LKGSFSEIPE LQVWYTKLGK TSERFLFKQL DSLWLLDSDG SFTLSLHEDE LFTLTTLTTG RKGSYPLPPK SQPFPSTYKD DFNVDYPFFS EAPNFADQTG VFEYFTNIED PGEHHFTLRQ VLNQRPITWA ADASNTISII GDYNWTNLTT KCDVYIETPD TGGVFIAGRV NKGGILIRSA RGIFFWIFAN GSYRVTGDLA GWIIYALGRV EVTAKKWYTL TLTIKGHFAS GMLNDKSLWT DIPVNFPKNG WAAIGTHSFE FAQFDNFRVE ATR
This gene encodes a lysosomal protein which hydrolyzes the galactose ester bonds of galactosylceramide, galactosylsphingosine, lactosylceramide, and monogalactosyldiglyceride. Mutations in this gene have been associated with Krabbe disease, also known as globoid cell leukodystrophy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3.2.1.46; beta-galactosidase; Galactocerebrosidase; Galactocerebroside beta-galactosidase; Galactosylceramidase; Galactosylceramide beta-galactosidase; galactosylceraminidase; GALCERase; testis tissue sperm-binding protein Li 88E; testis tissue sperm-binding protein Li 89A
Gene Aliases: GALC
UniProt ID: (Human) B7Z7Z2
Entrez Gene ID: (Human) 2581
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support