Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T.
Predicted reactivity: Mouse (88%), Rat (87%), Pig (90%), Bovine (90%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an enzyme that catalyzes the epimerization of hexose sugars such as glucose and galactose. The encoded protein is expressed in the cytoplasm and has a preference for galactose. The encoded protein may be required for normal galactose metabolism by maintaining the equilibrium of alpha and beta anomers of galactose.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aldose 1-epimerase; aldose mutarotase; epididymis secretory sperm binding protein Li 63p; galactomutarotase; galactose enzyme activator; Galactose mutarotase; galactose mutarotase (aldose 1-epimerase)
Gene Aliases: BLOCK25; GALM; GLAT; HEL-S-63p; IBD1
UniProt ID: (Human) Q96C23
Entrez Gene ID: (Human) 130589
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support