Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
Recommended positive controls: H1299, Neuro2A, C2C12, PC-12, Rat2.
Predicted reactivity: Mouse (89%), Rat (90%), Zebrafish (81%), Xenopus laevis (84%), Xenopus Tropicalis (80%), Dog (92%), Pig (90%), Bovine (90%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes N-acetylgalactosamine-6-sulfatase whichis a lysosomal exohydrolase required for the degradation of theglycosaminoglycans, keratan sulfate, and chondroitin 6-sulfate.Sequence alterations including point, missense and nonsensemutations, as well as those that affect splicing, result in adeficiency of this enzyme. Deficiencies of this enzyme lead toMorquio A syndrome, a lysosomal storage disorder.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chondroitinase; Chondroitinsulfatase; galactosamine (N-acetyl)-6-sulfate sulfatase; Galactose-6-sulfate sulfatase; GalN6S; GalNAc6S sulfatase; N-acetylgalactosamine-6-sulfatase; N-acetylgalactosamine-6-sulfate sulfatase
Gene Aliases: GalN6S; GALNAC6S; GALNS; GAS; mFLJ00319; MPS4A
UniProt ID: (Human) P34059, (Mouse) Q571E4, (Rat) Q32KJ6
Entrez Gene ID: (Human) 2588, (Mouse) 50917, (Rat) 292073
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support