Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Polypeptide N-acetylgalactosaminyltransferase that catalyzes the initiation of protein O-linked glycosylation and is involved in left/right asymmetry by mediating O-glycosylation of NOTCH1. O-glycosylation of NOTCH1 promotes activation of NOTCH1, modulating the balance between motile and immotile (sensory) cilia at the left-right organiser (LRO). Polypeptide N- acetylgalactosaminyltransferases catalyze the transfer of an N- acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor. Displays the same enzyme activity toward MUC1, MUC4, and EA2 than GALNT1. Not involved in glycosylation of erythropoietin (EPO).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GalNAc-T11; Polypeptide GalNAc transferase 11; Polypeptide N-acetylgalactosaminyltransferase 11; pp-GaNTase 11; Protein-UDP acetylgalactosaminyltransferase 11; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 11; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 11 (GalNAc-T11)
Gene Aliases: A430075I06Rik; AI648252; E430002F06Rik; GALNAC-T11; GALNACT11; GALNT11
UniProt ID: (Human) Q8NCW6, (Mouse) Q921L8, (Rat) Q6P6V1
Entrez Gene ID: (Human) 63917, (Mouse) 231050, (Rat) 311952
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support