Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins (see MUC3A, MIM 158371) by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.[supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16031; FLJ41157; GalNAc transferase 13; GalNAc-T13; H_NH0187G20.1; MGC119459; MGC119461; Polypeptide GalNAc transferase 13; Polypeptide N-acetylgalactosaminyltransferase 13; pp-GalNAc-T13; pp-GaNTase 13; Protein-UDP acetylgalactosaminyltransferase 13; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 13; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13 (GalNAc-T13); WUGSC:H_NH0187G20.1
Gene Aliases: A230002A12; A230020F20; BB182356; GalNAc-T13; GALNT13; KIAA1918; T13
UniProt ID: (Human) Q8IUC8, (Mouse) Q8CF93, (Rat) Q6UE39
Entrez Gene ID: (Human) 114805, (Mouse) 271786, (Rat) 311039
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support