Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Growth arrest-specific 7 is expressed primarily in terminally differentiated brain cells and predominantly in mature cerebellar Purkinje neurons. GAS7 plays a putative role in neuronal development. Several transcript variants encoding proteins which vary in the N-terminus have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GAS-7; growth arrest-specific 7; Growth arrest-specific protein 7; MLL/GAS7 fusion protein
Gene Aliases: GAS7; KIAA0394; MLL/GAS7
UniProt ID: (Human) O60861
Entrez Gene ID: (Human) 8522
Molecular Function: actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support