Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SHLFGFPPT PPKEVSPDPS TTGAASPASS SAGGSAARGE DKDGVKYQVS LTESMKMESG SPLRPGLATM GTQPATHHPI PTYPSYVPAA AHDYSSGLFH PGGFLGGPAS SFTPKQRSKA RSCSEGRECV NCGATATPLW RRDGTGHYLC NACGLYHKMN GQNRPLIKPK RRLTTTTTLW RRNANGDPVC NACGLYYKLH NVNRPLTMKK EGIQTRNRKM SNKSKKSKKG AECFEELSKC MQEKSSPFSA AALAGHMAPV GHLPPFSHSG HILPTPTPIH PSSSLSFGHP HPSSMVTAMG (168-467 aa encoded by BC018988)
GATA2 (GATA binding protein 2) belongs to the GATA family of transcription factors. GATA proteins play a very important role in signaling pathways controlling proliferation and differentiation of erythroid progenitor cells. There are 6 transcription factors in the GATA family: GATA1 to GATA6. GATA2 is a transcriptional activator regulating endothelin-1 gene expression in endothelial cells. It also appears to be involved in differentiation of other tissue types (uro-genital and nervous) during embryogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endothelial transcription factor GATA-2; FLJ45948; GATA-binding protein 2; GATA-bindning protein 2; gata2 {ECO:0000312|EMBL:AAK51128.1}; MGC2306
Gene Aliases: DCML; Gata-2; GATA2; IMD21; MONOMAC; NFE1B
UniProt ID: (Human) P23769, (Mouse) O09100, (Rat) Q924Y4
Entrez Gene ID: (Human) 2624, (Mouse) 14461, (Rat) 25159
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support