Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MEVAPEQPRW MAHPAVLNAQ HPDSHHPGLA HNYMEPAQLL PPDEVDVFFN HLDSQGNPYY ANPAHARARV SYSPAHARLT GGQMCRPHLL HSPGLPWLDG GK
GATA2 (GATA binding protein 2) belongs to the GATA family of transcription factors. GATA proteins play a very important role in signaling pathways controlling proliferation and differentiation of erythroid progenitor cells. There are 6 transcription factors in the GATA family: GATA1 to GATA6. GATA2 is a transcriptional activator regulating endothelin-1 gene expression in endothelial cells. It also appears to be involved in differentiation of other tissue types (uro-genital and nervous) during embryogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endothelial transcription factor GATA-2; FLJ45948; GATA-binding protein 2; MGC2306
Gene Aliases: DCML; GATA2; IMD21; MONOMAC; NFE1B
UniProt ID: (Human) P23769
Entrez Gene ID: (Human) 2624
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support