Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Immunogen sequence: STQAATASSR NSCAADDKAT EPLPKDCPVS SYNEWDPLEE VIVGRAENAC VPPFTIEVKA NTYEKYWPFY QKQGGHYFPK DHLKKAVAEI EEMCNILKTE GVTVRRPDPI DWSLKYKTPD FESTGLYSAM PRDILIVVGN EIIEAPMAWR SRFFEYRAYR SIIKDYFHRG AKWTTAPKPT MADELYNQDY PIHSVEDRHK LAAQGKFVTT EFEPCFDAAD FIRAGRDIFA QRSQVTNYLG IEWMRRHLAP DYRVHIISFK DPNPMHIDAT FNIIGPGIVL SNPDRPCHQI DLFKKAGWTI ITPPTPIIPD DHPLWMSSKW LSMNVLMLDE KRVMVDANEV PIQKMFEKLG ITTIKVNIRN ANSLGGGFHC WTCDVRRRGT LQSYLD
AGAT, also known as glycine amidinotransferase (L-arginine:glycine amidinotransferase), GATM or transamidinase, is a 423 amino acid protein belonging to the amidinotransferase family. Encoded by a gene that maps to human chromosome 15q21.1, AGAT exists as three alternatively spliced isoforms and consists of a homodimer, with equilibrium between monomeric and dimeric forms favoring a monomer subunit structure. AGAT localizes to mitochondrial inner membranes, peripheral membranes and cytoplasm. Biallelically expressed in placenta and fetal tissues, AGAT is also expressed in brain, heart, liver, lung, salivary gland and skeletal muscle tissue, with high expression in kidney. AGAT is elevated in the myocardium during heart failure and is decreased in inter-uterine growth restriction (IUGR)-associated placenta. AGAT catalyzes biosynthesis of guanidinoacetate, the immediate precursor of creatine, which plays a vital role in energy metabolism in muscle tissues. AGAT defects are associated with arginine:glycine amidinotransferase deficiency, an autosomal recessive disorder characterized by developmental delay or regression, mental retardation, severe disturbance of expressive and cognitive speech and severe depletion of creatine/phosphocreatine in brain. AGAT may be linked to embryonic and central nervous system development and may function in heart failure response by elevating local creatine synthesis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AT; glycine amidinotransferase (L-arginine:glycine amidinotransferase); Glycine amidinotransferase, mitochondrial; L-arginine: glycine amidinotransferase; L-arginine:glycine amidinotransferase; testicular secretory protein Li 19; Transamidinase
Gene Aliases: 1810003P21Rik; AGAT; AI314789; AT; CCDS3; GATM
UniProt ID: (Human) P50440, (Mouse) Q9D964, (Rat) P50442
Entrez Gene ID: (Human) 2628, (Mouse) 67092, (Rat) 81660
Molecular Function:
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support