Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAPEINLPGP MSLIDNTKGQ LVVNPEALKI LSAITQPVVV VAIVGLYRTG KSYLMNKLAG KKNGFSLGST VKSHTKGIWM WCVPHPKKPE HTLVLLDTEG LGDIEKGDNE NDSWIFALAI LLSSTFVYNS MGTINQQAMD QLHYVTELTD RIKANSSPGN NSVDDSADFV SFFPAFVWTL RDFTLELEVD GEPITADDYL ELSLKLRKGT DKKSKSFNDP RLCIRKFFPK RKCFVFDWPA PKKYLAHLEQ LKEEELNPDF IEQVAEFCSY ILSHSNVKTL SGGIPVNGPR LESLVLTYVN AISSGDLPCR ENAVLALAQI ENSAAVEKAI AHYEQQMGQK VQLPTETLQE LLDLHRDSER EAIEVFMKNS FKDVDQMFQR KLGAQLEARR DDFCKQNSKA SSDCCMALLQ DIFGPLEEDV KQGTFSKPGG YRLFTQKLQE LKNKYYQVPR KGIQAKEVLK KYLESKEDVA DALLQTDQSL SEKEKAIEVE RIKAESAEAA KKMLEEIQKK NEEMMEQKEK SYQEHVKQLT EKMERDRAQL MAEQEKTLAL KLQEQERLLK EGFENESKRL QKDIWDIQMR SKSLEPICNI L
Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GBP-2; GTP-binding protein 2; Guanine nucleotide-binding protein 2; guanylate binding protein 2, interferon-inducible; Guanylate-binding protein 2; Interferon-induced guanylate-binding protein 2
Gene Aliases: GBP2
UniProt ID: (Human) P32456
Entrez Gene ID: (Human) 2634
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support