Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GBP-2; GTP-binding protein 2; Guanine nucleotide-binding protein 2; guanylate binding protein 2, interferon-inducible; Guanylate-binding protein 2; Interferon-induced guanylate-binding protein 2
Gene Aliases: GBP2
UniProt ID: (Human) P32456
Entrez Gene ID: (Human) 2634
Molecular Function: heterotrimeric G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support