Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLEEQEKTLT SKLQEQARVL KERCQGESTQ LQNEIQKLQK TLKKKTKRYM SHK
Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 43%.
This gene encodes a member of the guanylate-binding protein (GBP) family. GBPs specifically bind guanine nucleotides (GMP, GDP, and GTP) and contain two of the three consensus motifs found in typical GTP-binding proteins. The encoded protein interacts with a member of the germinal center kinase family. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GBP-3; GTP-binding protein 3; Guanine nucleotide-binding protein 3; Guanylate-binding protein 3; guanylate-binding protein 3 delta C; RP4-531M19.1
Gene Aliases: GBP3
UniProt ID: (Human) Q9H0R5
Entrez Gene ID: (Human) 2635
Molecular Function: heterotrimeric G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support