Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: LPQAALQPA LPPAHPHHQI PSLPTGFCSS LAQGMALTST LMATLPGGFS ASPQHQEAAA ARKFAPQPLP GGGNFDKAEA LQADAEDGKG FLAKEGSLLA FSAAETVQAS LVGAVRGQGK DESKVEDDPK GKEESFSLES DVDYSSDDNL TGQAAHKEED PGHALEETPP SSGAAGSTTS TGKNR (65-248 aa encoded by B C137448)
Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gastrulation and brain-specific homeobox protein 2; Homeobox protein GBX-2; Stimulated by retinoic acid gene 7 protein
Gene Aliases: D130058E05Rik; Gbx-2; GBX2; MMoxA; Stra7
UniProt ID: (Human) P52951, (Mouse) P48031
Entrez Gene ID: (Human) 2637, (Mouse) 14472
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support