Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MPYLLISTQI RMEVGPTMVG DEQSDPELMQ HLGASKRRAL GNNFYEYYVD DPPRIVLDKL ERRGFRVLSM TGVGQTLVWC LHKE
Mediates tetrahydrobiopterin inhibition of GTP cyclohydrolase 1. This inhibition is reversed by L-phenylalanine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GFRP; GTP cyclohydrolase 1 feedback regulatory protein; GTP cyclohydrolase I feedback regulatory protein; p35
Gene Aliases: GCHFR; GFRP; HsT16933; P35
UniProt ID: (Human) P30047
Entrez Gene ID: (Human) 2644
Molecular Function:
protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support