Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: LNSYLENMEV DVDTRCSILN YLKLIKKRAS GELMTVARWM REFIANHPDY KQDSVITDEM NYSLILKCNQ IANELCECPE LLGSAFRKVK YSGSKTDSSN
Glutamate-cysteine ligase, also known as gamma-glutamylcysteine synthetase is the first rate-limiting enzyme of glutathione synthesis. The enzyme consists of two subunits, a heavy catalytic subunit and a light regulatory subunit. This locus encodes the catalytic subunit, while the regulatory subunit is derived from a different gene located on chromosome 1p22-p21. Mutations at this locus have been associated with hemolytic anemia due to deficiency of gamma-glutamylcysteine synthetase and susceptibility to myocardial infarction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gamma-ECS; Gamma-glutamylcysteine synthetase; GCS heavy chain; Glutamate--cysteine ligase catalytic subunit; Glutamylcysteine gamma synthetase light chain
Gene Aliases: Gclc; Glclc
UniProt ID: (Rat) P19468
Entrez Gene ID: (Rat) 25283
Molecular Function: ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support