Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MGTDSRAAKA LLARARTLHL QTGNLLNWGR LRKKCPSTHS EELHDCIQKT LNEWSSQINP DLVREFPDVL ECTVSHAVEK INPDEREEMK VSAKLFIVES NSSSSTRSAV DMACSVLGVA QLDSVIIASP PIEDGVNLSL EHLQPYWEEL ENLVQSKKIV AIGTSDLDKT QLEQLYQWAQ VKPNSNQVNL ASCCVMPPDL TAFAKQFDIQ LLTHNDPKEL LSGASFQEAL QESIPDIQAH EWVPLWLLRY SVIVKSRGII KSKGYILQAK RRGS
Gamma-glutamylcysteine synthetase (gamma-GCS) is the rate limiting enzyme for glutathione (L-gamma-glutamyl-L-cysteinylglycine, GSH) synthesis. GSH is ubiquitous in mammalian cells as a vital intra- and extracellular protective antioxidant. Gamma-GCS is a heterodimer of a heavy catalytic subunit and a light regulatory subunit that is responsive to inflammation, phenolic ntioxidants, heat shock, oxidants and cytokines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gamma glutamylcysteine synthetase; Gamma-ECS regulatory subunit; Gamma-glutamylcysteine synthetase regulatory subunit; GCS light chain; glutamate cysteine ligase (gamma-glutamylcysteine synthetase), regulatory; Glutamate--cysteine ligase modifier subunit; Glutamate--cysteine ligase regulatory subunit; glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory (30.8kD); glutamate-cysteine ligase modifier subunit; glutamate-cysteine ligase modifier subunit delta2 alternative splicing; glutamate-cysteine ligase regulatory protein; glutamate-cysteine ligase, modifier subunit; GSC light chain; null
Gene Aliases: GCLM; GLCLR
UniProt ID: (Human) P48507, (Rat) P48508
Entrez Gene ID: (Human) 2730, (Rat) 29739
Molecular Function:
ligase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support