Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Recommended positive controls: NIH-3T3.
Predicted reactivity: Mouse (97%), Zebrafish (90%), Xenopus laevis (90%), Rabbit (97%), Chimpanzee (100%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The gamma-Tubulin complex is composed of gamma Tubulin and the gamma-Tubulin complexassociated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the gamma-Tubulin complex. GCP4 (gamma-Tubulin complex component 4), also known as TUBGCP4, is a ubiquitously expressed 667 amino acid member of the gamma-Tubulin complex that localizes to the metaphase spindle during mitosis. In response to proteosome inhibition, GCP4 exhibits increased accumulation at the pericentiolar material where it participates in microtubule organization and nucleation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gamma tubulin ring complex protein; gamma tubulin ring complex protein (76p gene); Gamma-ring complex protein 76 kDa; Gamma-tubulin complex component 4; GCP-4; h76p; hGCP4; hGrip76; tubulin, gamma complex associated protein 4
Gene Aliases: 4932441P04Rik; 76P; D2Ertd435e; GCP-4; GCP4; Grip76; MCCRP3; TUBGCP4
UniProt ID: (Human) Q9UGJ1, (Mouse) Q9D4F8
Entrez Gene ID: (Human) 27229, (Mouse) 51885
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support