Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCS1 cleaves the distal alpha 1,2-linked glucose residue from the Glc(3)Man(9)GlcNAc(2) oligosaccharide precursor in a highly specific manner. Defects in GCS1 are the cause of type IIb congenital disorder of glycosylation (CDGIIb). This syndrome is also known as glucosidase I deficiency and is characterized by marked generalized hypotonia and hypomotility of the neonate, dysmorphic features, including a prominent occiput, short palpebral fissures, retrognathia, high arched palate, generalized edema, and hypoplastic genitalia. Symptoms include hepatomegaly, hypoventilation, feeding problems and seizures. The clinical course is progressive and survival is at most a few months.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endoplasmic reticulum alpha-glucosidase I; ER glu I; Glucosidase 1; glucosidase I; Glycoprotein-processing glucosidase I; Mannosyl-oligosaccharide glucosidase; Processing A-glucosidase I
Gene Aliases: 1810017N02Rik; AI181835; CDG2B; CWH41; DER7; GCS1; MOGS
UniProt ID: (Human) Q13724, (Mouse) Q80UM7
Entrez Gene ID: (Human) 7841, (Mouse) 57377
Molecular Function: glucosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support