Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Recommended positive controls: HeLa, HCT116.
Predicted reactivity: Mouse (88%), Rat (88%), Bovine (86%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytoplasmic PSD95 interactor; GAH; Guanase; guanine aminase; Guanine aminohydrolase; Guanine deaminase; p51-nedasin
Gene Aliases: AU015411; AW047581; CYPIN; GDA; GUANASE; KIAA1258; NEDASIN
UniProt ID: (Human) Q9Y2T3, (Mouse) Q9R111
Entrez Gene ID: (Human) 9615, (Mouse) 14544, (Rat) 83585
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support