Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MCAAQMPPLA HIFRGTFVHS TWTCPMEVLR DHLLGVSDSG KIVFLEEASQ QEKLAKEWCF KPCEIRELSH HEFFMPGLVD THIHASQYSF AGSSIDLPLL EWLTKYTFPA EHRFQNIDFA EEVYTRVVRR TLKNGTTTAC YFATIHTDSS LLLADITDKF GQRAFVGKVC MDLNDTFPEY KETTEESIKE TERFVSEMLQ KNYSRVKPIV TPRFSLSCSE TLMGELGNIA KTRDLHIQSH ISENRDEVEA VKNLYPSYKN YTSVYDKNNL LTNKTVMAHG CYLSAEELNV FHERGASIAH CPNSNLSLSS GFLNVLEVLK HEVKIGLGTD VAGGYSYSML DAIRRAVMVS NILLINKVNE KSLTLKEVFR LATLGGSQAL GLDGEIGNFE VGKEFDAILI NPKASDSPID LFYGDFFGDI SEAVIQKFLY LGDDRNIEEV YVGGKQVVPF SSSV
GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytoplasmic PSD95 interactor; GAH; Guanase; guanine aminase; Guanine aminohydrolase; Guanine deaminase; p51-nedasin
Gene Aliases: CYPIN; GDA; GUANASE; KIAA1258; NEDASIN
UniProt ID: (Human) Q9Y2T3
Entrez Gene ID: (Human) 9615
Molecular Function:
deaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support