Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SLC25A16 is a protein that contains three tandemly repeated mitochondrial carrier protein domains. The protein is localized in the inner membrane and facilitates the rapid transport and exchange of molecules between the cytosol and the mitochondrial matrix space. SLC25A16 gene has a possible role in Graves' disease.This gene encodes a protein that contains three tandemly repeated mitochondrial carrier protein domains. The encoded protein is localized in the inner membrane and facilitates the rapid transport and exchange of molecules between the cytosol and the mitochondrial matrix space. This gene has a possible role in Graves' disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDA; GDC; Graves disease autoantigen; Graves disease carrier protein; Graves' didease protein; hGP; MGC39851; Mitochondrial solute carrier protein homolog; solute carrier family 25 (mitochondrial carrier; Graves disease autoantigen), member 16; Solute carrier family 25 member 16
Gene Aliases: D10S105E; GDA; GDC; HGT.1; hML7; ML7; SLC25A16
UniProt ID: (Human) P16260
Entrez Gene ID: (Human) 8034
Molecular Function: mitochondrial carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support