Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDF-3; Growth/differentiation factor 3; VG-1-related protein 2
Gene Aliases: C78318; ecat9; Gdf-3; GDF3; KFS3; MCOP7; MCOPCB6; RGD1564178; UNQ222/PRO248; Vgr-2; Vgr2
UniProt ID: (Human) Q9NR23, (Mouse) Q07104
Entrez Gene ID: (Human) 9573, (Rat) 500311, (Mouse) 14562
Molecular Function: growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support