Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LGWDDWIIAP LDYEAYHCEG LCDFPLRSHL EPTNHAIIQT LLNSMAPDAA PASCCVPARL SPISILYIDA ANNVVYKQYE DMVVEACGCR
This gene encodes a member of the bone morphogenetic protein (BMP) family. BMPs belong to the transforming growth factor-beta superfamily of secreted signalling molecules that regulate diverse processes in growth, repair and embryonic development. In mouse, this gene functions as an inductive signal from the roof plate required for the specification of neuronal identity in the dorsal spinal cord.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDF-7; Growth/differentiation factor 7
Gene Aliases: BMP12; GDF7
UniProt ID: (Human) Q7Z4P5
Entrez Gene ID: (Human) 151449
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support