Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MDEEYDVIVL GTGLTECILS GIMSVNGKKV LHMDRNPYYG GESSSITPLE ELYKRFQLLE GPPESMGRGR DWNVDLIPKF LMANGQLVKM LLYTEVTRYL DFKVVEGSFV YKGGKIYKVP STETEALASN LMGMFEKRRF RKFLVFVANF DENDPKTFEG VDPQTTSMRD VYRKFDLGQD VIDFTGHALA LYRTDDYLDQ PCLETVNRIK LYSESLARYG KSPYLYPLYG LGELPQGFAR LSAIYGGTYM LNKPVDDIIM ENGKVVGVKS EGEVARCKQL ICDPSYIPDR VRKAGQVIRI ICILSHPIKN TNDANSCQII IPQNQVNRKS DIYVCMISYA HNVAAQGKYI AIASTTVETT DPEKEVEPAL ELLEPIDQKF VAISDLYEPI DDGCESQVFC SCSYDATTHF ETTCNDIKDI YKRMAGTAFD FENMKRKQND VFGEAEQ
GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI1 is expressed primarily in neural and sensory tissues. Mutations in GDI1 have been linked to X-linked nonspecific mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDI-1; Guanosine diphosphate dissociation inhibitor 1; mental retardation, X-linked 48; Oligophrenin-2; Protein XAP-4; Rab GDI alpha; Rab GDP dissociation inhibitor alpha; rab GDP-dissociation inhibitor, alpha; RABGD RABGDIA; testis secretory sperm-binding protein Li 208a
Gene Aliases: 1A; GDI1; GDIL; MRX41; MRX48; OPHN2; RABGD1A; RABGDIA; XAP-4; XAP4
UniProt ID: (Human) P31150
Entrez Gene ID: (Human) 2664
Molecular Function: G-protein modulator protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support