Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDEEYDVIV LGTGLTECIL SGIMSVNGKK VLHMDRNPYY GGESSSITPL EELYKRFQLL EGPPESMGRG RDWNVDLIPK FLMANGQLVK MLLYTEVTRY LDFKVVEGSF VYKGGKIYKV PSTETEALAS NLMGMFEKRR FRKFLVFVAN FDENDPKTFE GVDPQTTSMR DVYRKFDLGQ DVIDFTGHAL ALYRTDDYLD QPCLETVNRI (1-209 aa encoded by BC000317)
GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI1 is expressed primarily in neural and sensory tissues. Mutations in GDI1 have been linked to X-linked nonspecific mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDI-1; GDP-dissociation inhibitor 1; Guanosine diphosphate dissociation inhibitor 1; mental retardation, X-linked 48; Oligophrenin-2; Protein XAP-4; Rab GDI alpha; Rab GDIalpha; Rab GDP dissociation inhibitor alpha; rab GDP-dissociation inhibitor, alpha; RABGD RABGDIA; testis secretory sperm-binding protein Li 208a
Gene Aliases: 1A; GDI1; GDIA; GDIalpha; GDIL; MRX41; MRX48; OPHN2; RABGD1A; RABGDIA; XAP-4; XAP4
UniProt ID: (Human) P31150, (Mouse) P50396, (Rat) P50398
Entrez Gene ID: (Human) 2664, (Pig) 100623607, (Mouse) 14567, (Rat) 25183
Molecular Function:
G-protein modulator
protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support